Learn More
Tube Al-23, une extrémité fermée, dia. ext. : 20 mm, dia. int. de : 15mm, Thermo Scientific™
Marque: Thermo Scientific Alfa Aesar 030077.EH
Code nomenclature Nacres: NA.23
Informations supplémentaires : Poids : 0.01000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Tube | |
One end closed | |
600 mm |