missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ alpha COP Polyclonal Antibody
GREENER_CHOICE

Code produit 11591472
Change view
Click to view available options
Quantité:
100 μg
Conditionnement:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11591472 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11591472 Fournisseur Invitrogen™ Code fournisseur PA1067

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

PA1-067 detects coatomer-protein I alpha (COPI alpha) from human and rat cells. PA1-067 has been successfully used in Western blot and immunocytochemistry procedures. By Western blot, this antibody detects an ~140 kDa protein representing COP alpha from PC-12 cell extract. This antibody also detects an unidentified protein at ~100 kDa. Immunocytochemical staining of alpha-COP using PA1-067 results in alpha-COP localization to the Golgi complex and to distinct vesicular structures scattered throughout the cytoplasm. PA1-067 immunogen is a synthetic peptide corresponding to residues M(1) L T K F E T K S A R V K G L S F H P(19) C of rat alpha-COP. The PA1-067 immunizing peptide (Cat. # PEP-125) is available for use in neutralization and control experiments.

In eukaryotic cells, protein transport between the endoplasmic reticulum and Golgi compartments is mediated in part by non-clathrin-coated vesicular coat proteins (COPs). Seven coat proteins have been identified, and they represent subunits of a complex known as coatomer. The subunits are designated alpha-COP, beta-COP, beta-prime-COP, gamma-COP, delta-COP, epsilon-COP, and zeta-COP. The alpha-COP, encoded by COPA, shares high sequence similarity with RET1P, the alpha subunit of the coatomer complex in yeast. Also, the N-terminal 25 amino acids of alpha-COP encode the bioactive peptide, xenin, which stimulates exocrine pancreatic secretion and may act as a gastrointestinal hormone. Alternative splicing results in multiple splice forms encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Spécification

Antigène alpha COP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1mg/mL BSA and 0.05% sodium azide
Expression COPA
Numéro d’ordre du gène P53621
Alias de gène AILJK; alpha coat protein; alpha-coat protein; Alpha-COP; AU040324; coatomer protein complex subunit alpha; coatomer subunit alpha; COPA; HEPCOP; HEP-COP; proxenin; xenin; Xenopsin-related peptide
Symboles de gène(s) COPA
Espèces hôtes Rabbit
Immunogène Synthetic peptide corresponding to residues M(1) L T K F E T K S A R V K G L S F H P(19) C of rat alpha-COP.
Méthode de purification Antigen affinity chromatography
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1314, 304978
Espèces cibles Human, Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.