Learn More
Abnova™ AMBP (Human) Recombinant Protein
Human AMBP full-length ORF ( AAH41593, 19 a.a. - 352 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000259-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: AGPVPTPPDNIQVQGNFNISRIYGKWYNLAIGSTCPWLKKIMDRMTVSTLVLGEGATEAEISMTSTRWRKGVCEETSGAYEKTDTDGKFLYHKSKWNITMESYVVHTNYDEYAIFLTKKFSRHHGPTITAKLYGRAPQLRETLLQDFRVVAQGVGIPEDSIFTMADRGECVPGEQEPEPILIPRVRRAVLPQEEEGSGGGQLVTEVTKKEDSCQLGYSAGPCMGMTSRYFYNGTSMACETFQYGGCMGNGNNFVTEKECLQTCRTVAACNLPIVRGPCRAFIQLWAFDAVKGKCVLFPYGGCQGNGNKFYSEKECREYCGVPGDGDEELLRFSN
Spécification
AAH41593 | |
alpha-1-microglobulin/bikunin precursor | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AMBP | |
GST |
259 | |
Wheat germ expression system | |
25 μg | |
EDC1, HCP, HI30, IATIL, ITI, ITIL, ITILC, UTI | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |