Learn More
BMP5 (M82), Mouse anti-Human, Clone: 1G6, Abnova™
Description
This gene encodes a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. These proteins are synthesized as prepropeptides, cleaved, and then processed into dimeric proteins. This protein may act as an important signaling molecule within the trabecular meshwork and optic nerve head, and may play a potential role in glaucoma pathogenesis. This gene is differentially regulated during the formation of various tumors. [provided by RefSeq
Spécification
Spécification
| Antigène | BMP5 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1G6 |
| Conjugué | Unconjugated |
| Description | Mouse monoclonal antibody raised against a partial recombinant BMP5. |
| Formule | PBS with no preservative; pH 7.4 |
| Expression | BMP5 |
| Numéro d’ordre du gène | NM_021073.1 |
| Alias de gène | MGC34244 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.