Learn More
Caveolin 3 Mouse, Unlabeled, Clone: 26, BD
Code produit 15875449
Change view
Click to view available options
Quantité:
150 μg
50 μg
Conditionnement:
1 kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15875449 50 μg 1 kit
15885449 150 μg 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15875449 Fournisseur BD Biosciences Code fournisseur 610420
Mouse Monoclonal Antibody
Identified as a tyrosine phosphorylated protein in Rous sarcoma virus-transformed chick embryo fibroblasts (CEF), caveolin is now known to be ubiquitously expressed. Caveolin (also known as VIP21) localizes to non-clathrin membrane invaginations (caveolae) on the inner surface of the plasma membrane. This transmembrane protein plays a structural role in these specializations. Caveolin is also present at the trans-Golgi network (TGN), and similar quantities are found in apically and basolaterally destined transport vesicles. Caveolin is part of a complex containing glycosylphosphatidylinositol (GPI)-linked molecules and cytoplasmic signaling proteins. Caveolin is a transmembrane adaptor molecule that can simultaneously recognize GPI-linked proteins and interact with downstream cytoplasmic signaling molecules, such as c-yes, Annexin II, and hetero-trimeric G proteins. Caveolin 3 has been identified as a distinct isoform which is expressed only in smooth, skeletal, and cardiac muscle.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Caveolin 3
Applications Western Blot
Classification Monoclonal
Clone 26
Concentration 250μg/mL
Conjugué Unconjugated
Formule Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Espèces hôtes Mouse
Immunogène Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Méthode de purification Affinity Purified
Quantité 50 μg
État réglementaire RUO
Disciplines de recherche Cell Biology
Primaire ou secondaire Primary
Espèces cibles Mouse, Rabbit, Rat
Contenu et stockage Store undiluted at -20°C.
Isotype IgG1
Afficher plus Afficher moins
For Research Use Only.