Learn More
growth differentiation factor 10, Mouse, Polyclonal Antibody, Abnova™
Description
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis. [provided by RefSeq
Spécification
Spécification
| Antigène | growth differentiation factor 10 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugué | Unconjugated |
| Description | Mouse polyclonal antibody raised against a full-length recombinant GDF10. |
| Formule | 50% glycerol |
| Expression | GDF10 |
| Numéro d’ordre du gène | BC028237 |
| Alias de gène | BMP-3b/BMP3B |
| Symboles de gène(s) | GDF10 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.