Learn More
Abnova™ ANXA1 (Human) Recombinant Protein
Human ANXA1 full-length ORF ( AAH01275, 1 a.a. - 346 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000301-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: MAMVSEFLKQAWFIENEEQEYVQTVKSSKGGPGSAVSPYPTFNPSSDVAALHKAIMVKGVDEATIIDILTKRNNAQRQQIKAAYLQETGKPLDETLKKALTGHLEEVVLALLKTPAQFDADELRAAMKGLGTDEDTLIEILASRTNKEIRDINRVYREELKRDLAKDITSDTSGDFRNALLSLAKGDRSEDFGVNEDLADSDARALYEAGERRKGTDVNVFNTILTTRSYPQLRRVFQKYTKYSKHDMNKVLDLELKGDIEKCLTAIVKCATSKPAFFAEKLHQAMKGVGTRHKALIRIMVSRSEIDMNDIKAFYQKMYGISLCQAILDETKGDYEKILVALCGGN
Spécification
AAH01275 | |
annexin A1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ANXA1 | |
GST |
301 | |
Wheat germ expression system | |
25 μg | |
ANX1, LPC1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |