missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATF1 Monoclonal Antibody (ATF1 2A9/8)
Description
The predicted molecular weight of MA1-21313 is 60 kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes an activating transcription factor, which belongs to the ATF subfamily and bZIP (basic-region leucine zipper) family. It influences cellular physiologic processes by regulating the expression of downstream target genes, which are related to growth, survival, and other cellular activities. This protein is phosphorylated at serine 63 in its kinase-inducible domain by serine/threonine kinases, cAMP-dependent protein kinase A, calmodulin-dependent protein kinase I/II, mitogen- and stress-activated protein kinase and cyclin-dependent kinase 3 (cdk-3). Its phosphorylation enhances its transactivation and transcriptional activities, and enhances cell transformation. Fusion of this gene and FUS on chromosome 16 or EWSR1 on chromosome 22 induced by translocation generates chimeric proteins in angiomatoid fibrous histiocytoma and clear cell sarcoma. This gene has a pseudogene on chromosome 6.
Spécification
Spécification
| Antigène | ATF1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | ATF1 2A9/8 |
| Concentration | 0.7 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with no preservative; pH 7.2 |
| Expression | ATF1 |
| Numéro d’ordre du gène | P18846 |
| Alias de gène | AATase 1; Activating transcription factor 1; alcohol O-acetyltransferase; Alcohol O-acetyltransferase 1; ATF1; Atf-1; cAMP-dependent transcription factor ATF-1; cyclic AMP-dependent transcription factor ATF-1; EWS-ATF1; FUS/ATF-1; Protein TREB36; TCR-ATF1; TREB36; Unknown (protein for MGC:64099); wu:fc92c07; YOR377W; zgc:64099 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?