Learn More
Abnova™ AZGP1 (Human) Recombinant Protein
Human AZGP1 full-length ORF ( AAH33830, 22 a.a. - 298 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00000563-P01.25ug
Informations supplémentaires : Poids : 0.00010kg
Description
- Sequence: ENQGGRYSLTYIYTGLSKHVEDVPAFQALGSLNDLQFFRYNSKDRKSQPMGLWRQVEGMEDWKQDSQLQKAREDIFMETLKDIVEYYNDSNGSHVLQGRFGCEIENNRSSGAFWKYYYDGKDYIEFNKEIPAWVPFDPAAQITKQKWEAEPVYVQRAKAYLEEECPATLRKYLKYSKNILDRQDPPSVVVTSHQAPGEKKKLKCLAYDFYPGKIDVHWTRAGEVQEPELRGDVLHNGNGTYQSWVVVAVPPQDTAPYSCHVQHSSLAQPLVGPWEAS
Spécification
AAH33830 | |
alpha-2-glycoprotein 1, zinc-binding | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AZGP1 | |
GST |
563 | |
Wheat germ expression system | |
25 μg | |
ZA2G, ZAG | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |