Learn More
BD Human Th17/Treg Phenotyping Kit
Simultaneously view TH17 and regulatory T cells in human model systems
Marque:  BD 560762
Code nomenclature Nacres: NA.41
Code produit. 15870469
  • ONLINE EXCLUSIVE PRICE
    1000.00€
    995.10€ / 1 pièce
    Économisez 4.90€
    0.49% de réduction
Includes: Human Th17/Treg Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, FoxP3 Alexa Fluor™ 647), Human FoxP3 Buffer A (10X), Human FoxP3 Buffer B (50X), BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Allow for staining of surface CD4 along with intracellular factors
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor CD4+ T cell differentiation by cells cultured within various experimental model systems
TRUSTED_SUSTAINABILITY
Spécification
Spécification
1 Kit
For research use only. Not for diagnostic or therapeutic procedures.