Learn More
BD Mouse Th1/Th17 Phenotyping Kit
Simultaneously view TH17 and TH1 in mouse model systems
Marque:  BD 560759
Code nomenclature Nacres: NA.41
Code produit. 15850469
  • ONLINE EXCLUSIVE PRICE
    715.00€
    711.55€ / 1 pièce
    Économisez 3.45€
    0.48% de réduction
Includes: Mouse Th1/Th17 Phenotyping Cocktail (CD4 PerCP-Cy™ 5.5, IL-17A PE, Mouse IFN-γ FITC), BD Cytofix™ Fixation Buffer, BD Perm/Wash™ Buffer, BD GolgiStop™ Protein Transport Inhibitor
Description
Description
  • Provide a simple solution for measurement of multiple cytokines or transcriptions factors from a single sample using multicolor flow cytometry
  • Can be used to analyze ex vivo lymphoid cell samples or to monitor T helper cell differentiation
  • Ensures indentification and characterization of the nature of the T helper cell types
  • Allow for staining of surface CD4 along with intracellular factors
TRUSTED_SUSTAINABILITY
Spécification
Spécification
1 Kit
For research use only. Not for diagnostic or therapeutic procedures.