Learn More
R&D Systems™ Beclin 1 Antibody Pack
Code produit 18257623 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
3 Vials
Conditionnement:
4 pièces
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18257623 3 Vials 4 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 18257623 Fournisseur R&D Systems™ Code fournisseur NB91095610
 Article restreint
Expédition estimée: 06-11-2026
Includes
  • 1.NB110-87318: Beclin 1/ATG6 Antibody
  • 2.NB500-249: Beclin 1/ATG6 Antibody
  • 3.NB500-266: Beclin 1/ATG6 Antibody
A Novus product, part of R&D Systems. For Research applications
Beclin 1 specifically detects Beclin 1 in Human, Mouse, Rat samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot.
TRUSTED_SUSTAINABILITY
Spécification
Antigène Beclin 1
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Simple Western, Western Blot, KnockDown
Conjugué Unconjugated
Dilution Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated
Formule See individual datasheets
Alias de gène ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Symboles de gène(s) BECN1
Immunogène See individual datasheets
Quantité 3 Vials
Disciplines de recherche Cancer, mTOR Pathway
Identification génétique (Entrez) 8678
Comprend 1.NB110-87318: Beclin 1/ATG6 Antibody
2.NB500-249: Beclin 1/ATG6 Antibody
3.NB500-266: Beclin 1/ATG6 Antibody
Espèces cibles Human, Mouse, Rat
Contenu et stockage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Gamme de produits Novus