missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ beta COP Polyclonal Antibody
GREENER_CHOICE

Code produit 11511472
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
11511472 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11511472 Fournisseur Invitrogen™ Code fournisseur PA1061

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

PA1-061 detects Golgi beta-coatomer protein (beta-COP) from mouse, human and rat tissues. PA1-061 has been successfully used in Western blot and immunofluorescence procedures. By Western blot, this antibody detects an ~110kDa protein which represents beta-COP from rat brain homogenate. Indirect immunofluorescence staining of beta-COP in NIH-3T3 cells with PA1-061 results in beta-COP localization to the Golgi complex and to distinct vesicular structures scattered throughout the cytoplasm. Golgi localization of beta-COP is sensitive to treatment with brefeldin A or ATP depletion. The PA1-061 immunogen is a synthetic peptide corresponding to residues E(496) A G E L K P E E E I T V G P V Q K(513) of rat beta-COP. This sequence is completely conserved in human beta-COP.

This gene encodes a protein subunit of the coatomer complex associated with non-clathrin coated vesicles. The coatomer complex, also known as the coat protein complex 1, forms in the cytoplasm and is recruited to the Golgi by activated guanosine triphosphatases. Once at the Golgi membrane, the coatomer complex may assist in the movement of protein and lipid components back to the endoplasmic reticulum. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Spécification

Antigène beta COP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1mg/mL BSA and 0.05% sodium azide
Expression Copb1
Numéro d’ordre du gène P23514, P53618, Q9JIF7
Alias de gène 2610019B04Rik; beta coat protein; beta-coat protein; Beta-COP; coatomer protein complex subunit beta 1; coatomer protein complex, subunit beta 1; Coatomer subunit beta; COPB; COPB1; DKFZp761K102; FLJ10341; FLJ46444; FLJ57957; I79_013776; MSTP026; Rack2; receptor for activated C-kinase-2
Symboles de gène(s) Copb1
Espèces hôtes Rabbit
Immunogène Synthetic peptide corresponding to residues E(496) A G E L K P E E E I T V G P V Q K(513) of rat beta-COP.
Méthode de purification Ammonium sulfate precipitation
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 114023, 1315, 70349
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.