missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ beta Tubulin Monoclonal Antibody (1B12)
Description
beta Tubulin Monoclonal Antibody for Western Blot, ICC/IF
Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules. Beta-III tubulin is primarily expressed in neurons and may be involved in neurogenesis, axon guidance and maintenance. Mutations in the beta tubulin gene are the cause of congenital fibrosis of the extraocular muscles type 3. Beta-III tubulin was also detected in Sertoli cells of the testis and transiently in non-neuronal embryonic tissues. Glutamate residues at the C-terminus of beta III tubulin can be glutamylated. The precise function of such modifications is unclear. Tubulin is phosphorylated on Ser-172 by CDK1 during cell cycle progression. Ser-172 phosphorylation inhibits tubulin incorporation into microtubules.Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55 kDa subunits: alpha and Beta. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton. Because beta-tubulin is ubiquitously expressed in all eukaryotic cells, it is frequently used as a loading control for assays involving protein detection, such as Western blotting.
Spécification
Spécification
| Antigène | beta Tubulin |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 1B12 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 5mM sodium azide |
| Expression | Tubb4a |
| Numéro d’ordre du gène | A2AQ07, P04350, P07437, P69897, P85108, P99024, Q13509, Q13885, Q3KRE8, Q3ZCM7, Q4QRB4, Q767L7, Q7TMM9, Q922F4, Q9BUF5, Q9BVA1, Q9CWF2, Q9D6F9, Q9ERD7, Q9H4B7 |
| Alias de gène | 2310057H16Rik; 2810484G07Rik; 3200002H15Rik; AA408537; AI325297; AI596182; B130022C14Rik; bA631M21.2; BB220206; beta 5-tubulin; beta Ib tubulin; beta tubulin; beta tubulin 1; beta-4; beta-4 tubulin; beta-4-tubulin; beta-5; beta-5 tubulin; beta5-tubulin; betaIII-tubulin; beta-tubulin; beta-tubulin class-III; Beta-tubulin class-V; beta-tubulin class-VI; Beta-tubulin isotype I; c(beta)7 tubulin; CDCBM; CDCBM1; CDCBM5; CDCBM6; CFEOM3; CFEOM3A; class I beta tubulin; Class I beta-tubulin; class IIa beta-tubulin; class III beta-tubulin; class IVa beta-tubulin; class V beta tubulin; class V beta-tubulin; CSCSC1; dJ40E16.7; dystonia 4, torsion (autosomal dominant); DYT4; FEOM3; HSA10p15 beta-tubulin 4Q; HsT1601; hypothetical protein LOC540236; hypothetical protein LOC550458; I79_000194; I79_012097; LOC100349792; LOC100761889; LOW QUALITY PROTEIN: tubulin beta-6 chain; M(beta)1; M(beta)2; M(beta)3; M(beta)4; M(beta)5; M(beta)6; M40; M40 antibody; MGC128415 protein; MGC137802 protein; neuron-specific class III beta-tubulin; OK/SW-cl.56; OOMD; OOMD2; RGD1305887; si:ch211-122i22.4; si:ch211-9a18.2; Tubb; tubb beta-5; tubb.L; Tubb1; TUBB2; Tubb2a; tubb2a {ECO:0000250; Tubb3; TUBB4; TUBB4A; tubb5; TUBB-5; TUBB6; TUBB8; Tubulin 5 beta; tubulin beta; tubulin beta 1 class VI; tubulin beta 2A class IIa; tubulin beta 3 class III; tubulin beta 4'; tubulin beta 4A class IVa; tubulin beta 6 class V; tubulin beta 8 class VIII; Tubulin beta chain; tubulin beta class I; tubulin beta class I L homeolog; Tubulin beta class IIa; Tubulin beta class V; tubulin beta MGC4083; tubulin beta-1; tubulin beta-1 chain; tubulin beta-1 chain-like; tubulin beta-2A chain; tubulin beta-2B chain; tubulin beta-3 chain; Tubulin beta-4 chain; tubulin beta-4A chain; Tubulin beta-5 chain; tubulin beta-5 chain; tubulin beta-6 chain; tubulin beta-6 chain; Tubulin beta-7 chain; tubulin beta-8 chain; Tubulin beta-III; tubulin, beta 1 class VI; tubulin, beta 2; tubulin, beta 2A; tubulin, beta 2A class IIA; tubulin, beta 3; tubulin, beta 3 class III; tubulin, beta 4; tubulin, beta 4 class IVa; tubulin, beta 4A class IVa; tubulin, beta 5; tubulin, beta 5 class I; tubulin, beta 6 class V; tubulin, beta class I; tubulin, beta family 1; tubulin, beta polypeptide; tubulin, beta polypeptide 2; tubulin, beta, 3; tubulin, beta, 4; tubulin, beta, 5; uncharacterized protein LOC550458; UniProtKB:Q7TMM9}; Unknown (protein for MGC:134115); unnamed protein product; XELAEV_18039222mg; XLOT; zbetaI; zgc:112515 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?