Learn More
ACROBiosystems Biotinylated Cynomolgus CD24 Protein, Fc Tag, ultra sensitivity (primary amine labeling)
423.38€ - 1862.87€
Specifications
Poids moléculaire | 29.3 kDa |
---|---|
Source | Biotinylated Cynomolgus CD24, Fc Tag, primary amine labeling (CD4-C8254) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1). |
Immunogène | Thr 27 - Lys 54 |
Conditions de stockage | -20°C |
Alias de gène | CD24,CD24A |
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantité | Price | Quantity & Availability | |||||
17256100
|
ACROBiosystems
CD4-C8254-25UG |
25 μg |
423.38€
25µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17246100
|
ACROBiosystems
CD4-C8254-200UG |
200 μg |
1862.87€
200µg |
Expédition estimée: 30-04-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CD24 may have a pivotal role in cell differentiation of different cell types. Signaling could be triggered by the binding of a lectin-like ligand to the CD24 carbohydrates, and transduced by the release of second messengers derived from the GPI-anchor. Modulates B-cell activation responses. Promotes AG-dependent proliferation of B-cells, and prevents their terminal differentiation into antibody-forming cells. In association with SIGLEC10 may be involved in the selective suppression of the immune response to danger-associated molecular patterns (DAMPs) such as HMGB1, HSP70 and HSP90. Plays a role in the control of autoimmunity.
This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 29.3 kDa. The protein migrates as 45-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
29.3 kDa | |
Thr 27 - Lys 54 | |
CD24,CD24A | |
UltraLys | |
Fc Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |
Biotinylated Cynomolgus CD24, Fc Tag, primary amine labeling (CD4-C8254) is expressed from human 293 cells (HEK293). It contains AA Thr 27 - Lys 54 (Accession # I7GKK1-1). | |
-20°C | |
CD24 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |