Learn More
ACROBiosystems Biotinylated Human B7-H3 / CD276 Protein, Avitag™,His Tag
Biotinylated Protein
Marque: ACROBiosystems B73-H82E6-200UG
Informations supplémentaires : Poids : 0.00001kg
Description
B7 homolog 3 (B7-H3), a member of the immunoglobulin superfamily, is also known CD276, which contains two Ig-like C2-type (immunoglobulin-like) domains and two Ig-like V-type (immunoglobulin-like) domains. B7-H3 may participate in the regulation of T-cell-mediated immune response. B7-H3 also plays a protective role in tumor cells by inhibiting natural-killer mediated cell lysis as well as a role of marker for detection of neuroblastoma cells. Furthermore, B7-H3 is involved in the development of acute and chronic transplant rejection and in the regulation of lymphocytic activity at mucosal surfaces. It could also play a key role in providing the placenta and fetus with a suitable immunological environment throughout pregnancy.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 26.5 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_079516.1 | |
26.5 kDa | |
200 µg | |
Leu 29 - Pro 245 | |
B7-H3,CD276,B7 homolog 3 | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Biotinylated Human B7-H3, Avitag,His Tag (B73-H82E6) is expressed from human 293 cells (HEK293). It contains AA Leu 29 - Pro 245 (Accession # Q5ZPR3-2). | |
-20°C | |
B7-H3 | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |