Learn More
ACROBiosystems Biotinylated Human CD4 Protein, His, Avitag™
Biotinylated Protein
Marque: ACROBiosystems CD4-H82E8-25UG
Informations supplémentaires : Poids : 0.00001kg
Description
T-cell surface glycoprotein CD4 is also known as T-cell surface antigen T4/Leu-3. CD4 contains three Ig-like C2-type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. CD4 is accessory protein for MHC class-II antigen/T-cell receptor interaction. CD4 induces the aggregation of lipid rafts. CD4 is a primary receptor used by HIV-1 to gain entry into host T cells. HIV infection leads to a progressive reduction of the number of T cells possessing CD4 receptors. Therefore, medical professionals refer to the CD4 count to decide when to begin treatment for HIV-infected patients.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 45.0 kDa. The protein migrates as 55-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_000607.1 | |
45.0 kDa | |
25 µg | |
Lys 26 - Pro 396 | |
CD4,CD4mut,LEU3 | |
Precision Avi | |
PBS, pH7.4 | |
HEK293 | |
Human |
Powder | |
>95% as determined by SDS-PAGE | |
Biotinylated Human CD4, His,Avitag (CD4-H82E8) is expressed from human 293 cells (HEK293). It contains AA Lys 26 - Pro 396 (Accession # P01730-1). | |
-20°C | |
CD4 | |
His Tag and Avi Tag | |
Less than 1.0 EU / µg by the LAL method. | |
MABSol™ Biotin Labeled Proteins |