Learn More
ACROBiosystems Biotinylated Human EGFRvIII Protein, Avitag™,His Tag
MALS verified
447.57€ - 1947.54€
Spécification
Numéro d’adhésion | NP_001333870.1 |
---|---|
Poids moléculaire | 42.2 kDa |
Source | Biotinylated Human EGFRvIII, Avitag,His Tag (EGR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1). |
Immunogène | Leu 25 - Ser 378 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15876627
|
ACROBiosystems
EGR-H82E0-25UG |
25 μg |
447.57€
25µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15866627
|
ACROBiosystems
EGR-H82E0-200UG |
200 μg |
1947.54€
200µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.The type III EGF deletion-mutant receptor (EGFRvIII) is the most common mutation and was first identified in primary human glioblastoma tumors; EGFR gene amplification is correlated with the structural rearrangement of the gene. The EGFRvIII gene has an in-frame deletion of 801 base pairs, corresponding to exons 2–7 in the mRNA, resulting in the deletion of amino acids 30-297 in the extracellular domain and the generation of a glycine at the fusion point.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 42.2 kDa. The protein migrates as 60-90 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_001333870.1 | |
Biotinylated Human EGFRvIII, Avitag,His Tag (EGR-H82E0) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 378 (Accession # NP_001333870.1). | |
-20°C | |
EGFRvIII | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
42.2 kDa | |
Leu 25 - Ser 378 | |
EGFRvIII | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |