Learn More
ACROBiosystems Biotinylated Human IL-6 Protein, epitope tag free, ultra sensitivity (primary amine labeling)
350.80€ - 1681.42€
Spécification
Numéro d’adhésion | NP_000591 |
---|---|
Poids moléculaire | 20.8 kDa |
Source | MABSol™ Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It contains AA Val 30 - Met 212 (Accession # NP_000591). It is the biotinylated form of ActiveMax™ Human IL-6, Tag Free (IL6-H4218). |
Immunogène | Val 30 - Met 212 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15867057
|
ACROBiosystems
IL6-H8218-200UG |
200 μg |
1681.42€
200µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15877057
|
ACROBiosystems
IL6-H8218-25UG |
25 μg |
350.80€
25µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Interleukin 6 (IL-6) is also known as HGF, BSF2,HSF, IFNB2 and IL-6, originally identified as a B cell differentiation factor, is a multifunctional cytokine that regulates immune responses, hematopoiesis, acute phase responses, and inflammatory reactions.It is secreted by T cells, macrophages , monocytes, fibroblasts,endothelial cells,et.al. to stimulate immune response to trauma, especially burns or other tissue damage leading to inflammation. Interleukin 6 has been shown to interact with interleukin-6 receptor and glycoprotein. IL-6 is relevant to many disease processes such as diabetes,atherosclerosis, depression,Alzheimer's Disease,systemic,lupus erythematosus,prostate cancer and rheumatoid arthritis. Advanced/metastatic cancer patients have higher levels of IL-6 in their blood.Hence there is an interest in developing anti-IL-6 agents as therapy against many of these diseases.
The product does NOT contain any epitope tags. The protein has a calculated MW of 20.8 kDa. The protein migrates as 24-27 kDa on a SDS-PAGE gel under reducing (R) condition due to different glycosylation.Spécification
NP_000591 | |
MABSol™ Biotinylated Human IL-6, epitope tag free, primary amine labeling (IL6-H8218) is expressed from human HEK293 cells. It contains AA Val 30 - Met 212 (Accession # NP_000591). It is the biotinylated form of ActiveMax™ Human IL-6, Tag Free (IL6-H4218). | |
-20°C | |
IL-6 | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
20.8 kDa | |
Val 30 - Met 212 | |
IL6,Interleukin-6,BSF2,HSF,IFNB2 | |
UltraLys | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |