missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated Human PSCA Protein, His, Avitag (MALS verified)
423.38€ - 1862.87€
Spécification
Numéro d’adhésion | NP_005663.2 |
---|---|
Poids moléculaire | 12.1 kDa |
Source | Biotinylated Human PSCA, His,Avitag (PSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1). |
Immunogène | Leu 12 - Ser 86 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17256420
|
ACROBiosystems
PSA-H82E3-25UG |
25 μg |
423.38€
25µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17246420
|
ACROBiosystems
PSA-H82E3-200UG |
200 μg |
1862.87€
200µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
The Prostate stem cell antigen (PSCA) is a glycosylphosphatidylinositol (GPI)-anchored protein, plays an important role in tumorigenesis. The prostate stem cell antigen (PSCA) gene, which encodes a prostate-specific antigen (PSA), was identified as a gene involved in cell adhesion and proliferation. PSCA may be involved in the regulation of cell proliferation. Has a cell-proliferation inhibition activity in vitro. May act as a modulator of nicotinic acetylcholine receptors (nAChRs) activity. In vitro inhibits nicotine-induced signaling probably implicating alpha-3:beta-2- or alpha-7-containing nAChRs.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 12.1 kDa. The protein migrates as 20-30 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Spécification
NP_005663.2 | |
Biotinylated Human PSCA, His,Avitag (PSA-H82E3) is expressed from human 293 cells (HEK293). It contains AA Leu 12 - Ser 86 (Accession # O43653-1). | |
-20°C | |
PSCA | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
12.1 kDa | |
Leu 12 - Ser 86 | |
PSCA,UNQ206,PRO232 | |
Precision Avi | |
His Tag & Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>90% as determined by SDS-PAGE. |