Learn More
ACROBiosystems Biotinylated Human TNF-alpha Protein, epitope tag free, ultra sensitivity (primary amine labeling) (active trimer)
MALS verified
411.28€ - 1850.77€
Spécification
Numéro d’adhésion | NP_000585.2 |
---|---|
Poids moléculaire | 17.4 kDa |
Source | MABSol™ Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expressed from human HEK293 cells. It contains AA Val 77 - Leu 233 (Accession # NP_000585.2). It is the biotinylated form of ActiveMax™ Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211). |
Immunogène | Val 77 - Leu 233 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
15877837
|
ACROBiosystems
TNA-H8211-25UG |
25 μg |
411.28€
25µg |
Expédition estimée: 27-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
15867837
|
ACROBiosystems
TNA-H8211-200UG |
200 μg |
1850.77€
200µg |
Expédition estimée: 31-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
Tumor necrosis factor alpha (TNFα) is a cytokine produced primarily by monocytes and macrophages. It is found in synovial cells and macrophages in the tissues.The primary role of TNFα is in the regulation of immune cells. TNFα is able to induce apoptotic cell death, to induce inflammation, and to inhibit tumorigenesis and viral replication. Dysregulation of TNFα production has been implicated in a variety of human diseases, including major depression, Alzheimer's disease and cancer. Recombinant TNFα is used as an immunostimulant under the INN tasonermin. TNFα can be produced ectopically in the setting of malignancy and parallels parathyroid hormone both in causing secondary hypercalcemia and in the cancers with which excessive production is associated.
The product does NOT contain any epitope tags. The protein has a calculated MW of 17.4 kDa. The protein migrates as 18 kDa on a SDS-PAGE gel under reducing (R) condition.Spécification
NP_000585.2 | |
MABSol™ Biotinylated Human TNF-alpha, epitope tag free, primary amine labeling (active trimer) (MALS verified) (TNA-H8211) is expressed from human HEK293 cells. It contains AA Val 77 - Leu 233 (Accession # NP_000585.2). It is the biotinylated form of ActiveMax™ Human TNF-alpha, Tag Free, low endotoxin (active trimer) (MALS verified) (TNA-H4211). | |
-20°C | |
TNF-alpha | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
17.4 kDa | |
Val 77 - Leu 233 | |
DIF,TNF-alpha,TNFA,TNFSF2,cachexin,cachectin,TNFα | |
UltraLys | |
Tag Free | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by reduced SDS-PAGE |