Learn More
ACROBiosystems Biotinylated Rat CD3 epsilon&CD3 gamma Heterodimer Protein, His, Avitag&Flag Tag (MALS verified)
Marque: ACROBiosystems CDG-R82D4-25ug
Informations supplémentaires : Poids : 0.00010kg
Description
T-cell surface glycoprotein CD3 delta & CD3 gamma chain, also known as CD3D & CD3G or CD3D&CD3G respectively, are single-pass type I membrane proteins. CD3D, together with CD3- epsilon(CD3E) , CD3-gamma and CD3-zeta, and the T-cell receptor alpha/beta and gamma/delta heterodimers, forms the T cell receptor-CD3 complex. T cell receptor-CD3 complex plays an important role in coupling antigen recognition to several intracellular signal-transduction pathways.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 16.7 kDa | 15.5 kDa.Spécification
NP_001101610.1 (CD3E) & NP_001071114 (CD3G) | |
16.7 kDa, 15.5 kDa | |
Biotinylated Rat CD3E&CD3G; Heterodimer Protein, His, Avitag &Flag; Tag (CDG-R82D4) is expressed from human 293 cells (HEK293). It contains AA Gln 21 - Asp 103(CD3E) & Gln 23 - Ser 116 (CD3G) (Accession # NP_001101610.1 (CD3E) & NP_001071114 (CD3G)). | |
-20°C | |
CD3E & CD3G | |
His Tag & Avi Tag & Flag Tag | |
MABSol™ Biotin Labeled Proteins |
Powder | |
25 μg | |
Gln 21 - Asp 103(CD3E) & Gln 23 - Ser 116 (CD3G) | |
CD3 epsilon & CD3 gamma,CD3E & CD3G | |
Precision Avi | |
HEK293 | |
Rat |