missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Biotinylated SARS-CoV-2 S protein RBD (N501Y), His, Avitag
459.67€ - 2116.90€
Spécification
Numéro d’adhésion | QHD43416.1 (N501Y) |
---|---|
Poids moléculaire | 28.2 kDa |
Source | Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (SPD-C82E6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)). |
Immunogène | Arg 319 - Lys 537 |
Conditions de stockage | -20°C |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
17246540
|
ACROBiosystems
SPD-C82E6-200UG |
200 μg |
2116.90€
200µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
17256540
|
ACROBiosystems
SPD-C82E6-25UG |
25 μg |
459.67€
25µg |
Expédition estimée: 10-05-2024 Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
It's been reported that Coronavirus can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This protein carries a polyhistidine tag at the C-terminus, followed by an Avi tag. The protein has a calculated MW of 28.2 kDa.Spécification
QHD43416.1 (N501Y) | |
Biotinylated SARS-CoV-2 S protein RBD (N501Y), His,Avitag (SPD-C82E6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (N501Y)). | |
-20°C | |
S Protein RBD | |
SARS-CoV-2 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |
28.2 kDa | |
Arg 319 - Lys 537 | |
Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD,COVID-19 | |
Precision Avi | |
His Tag & Avi Tag | |
Powder |