Learn More
Invitrogen™ BITE Polyclonal Antibody
Code produit 16307284
Modifier l'affichage
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
16307284 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 16307284 Fournisseur Invitrogen™ Code fournisseur PA578585
 Article restreint
Expédition estimée: 10-12-2026
Rabbit Polyclonal Antibody
Positive Control: NT2D1 Predicted Reactivity: Rhesus Monkey (95%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
CEP70 plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, it is required for the organization and orientation of the mitotic spindle.
TRUSTED_SUSTAINABILITY
Spécification
Antigène BITE
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.01 mg/mL
Conjugué Unconjugated
Formule 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Expression CEP70
Numéro d’ordre du gène Q8NHQ1
Alias de gène 6720484E09Rik; BITE; C030018L16Rik; centrosomal protein 70; centrosomal protein 70kDa; Centrosomal protein of 70 kDa; CEP70; I79_009119; p10-binding protein; testicular tissue protein Li 36
Symboles de gène(s) CEP70
Espèces hôtes Rabbit
Immunogène Recombinant protein encompassing a sequence within the N-terminus region of human CEP70. The exact sequence is proprietary.
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 80321
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG