Learn More
Complexe de sulfure de borane-diméthyle, conditionné sous Argon dans des flacons ChemSeal™ refermables, Thermo Scientific Chemicals
Catalog Number 11380248
Click to view available options
Quantité:
5 mL
25 mL
100 mL
500 mL
13292-87-0
C2H9BS
75.96
MFCD00013189
RMHDLBZYPISZOI-UHFFFAOYSA-N
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
9833925
(methylsulfanyl)methane borane
B.CSC
13292-87-0
C2H9BS
75.96
MFCD00013189
RMHDLBZYPISZOI-UHFFFAOYSA-N
dimethyl sulfide borane, borane-methyl sulfide complex, borane dimethyl sulfide complex, borane-dimethyl sulfide, dimethyl sulfideborane, dimethyl sulfide trihydroboron, borane-dimethyl sulfide complex, borane methylsulfide, borane dimethylsulfide, borane dms
9833925
(methylsulfanyl)methane borane
B.CSC
Borane- dimethyl sulfide is used for hydroboration and reduction reactions. It is used in the reduction of aldehydes, ketones, epoxides and carboxylic acids to give corresponding alcohols. It is also involved in the Corey-Itsuno reduction. It acts as an intermediate in the preparation of highly pure semiconductor. It is also employed as a rocket propellant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Conditionnés sous Argon
TRUSTED_SUSTAINABILITY
Nom chimique ou matériau Borane-dimethyl sulfide complex
Point de fusion -38°C
Couleur Incolore à jaune
Densité 0.801 g/mL
Point d’ébullition ∼44°C (decomposition)
Point d’éclair −1°C (31°F)
Odeur Unpleasant
Conditionnement Flacons Chemseal™
Formule linéaire (CH3)2S·BH3
Indice de réfraction 1.457
Quantité 100 mL
Numéro UN UN2924
Beilstein 3663489
Sensibilité Air and moisture sensitive
Informations sur la solubilité Miscible with ethyl ether,tetrahydrofurane,dichloromethane,benzene,xylene,hexane,diglyme,dimethyl ether,ethyl acetate,toluene,methylene chloride and other aprotic solvents.
Poids de la formule 75.97
Forme physique Liquide