missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C8orf48 Polyclonal Antibody
GREENER_CHOICE

Code produit 15773531 Tous les produits Thermo Scientific Produits
Modifier l'affichage
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15773531 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15773531 Fournisseur Invitrogen™ Code fournisseur PA555614

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: RDAFSCTVPD ELLNRIYFKN MRTTPKQEAA AKQHISYQCP YCNRKRAELA LSAFLKQKKT LLESFLLQER IDEHLHT Highest antigen sequence identity to the following orthologs: Mouse - 66%, Rat - 25%.

The C8orf48 gene has been identified as a critical biomarker in colorectal cancer (CRC). Research indicates that C8orf48 plays a significant role in inhibiting tumorigenesis by regulating key signaling pathways. This gene is negatively associated with KRAS, BRAF, and MAP2K2 in normal colorectal tissues and negatively correlated with MAP2K2 in CRC tissues. These associations suggest that C8orf48 may function as a tumor suppressor by modulating pathways involved in cell proliferation, migration, and invasion. The gene's expression and methylation profiles have been extensively studied using data from The Cancer Genome Atlas (TCGA), and functional assays have confirmed its inhibitory effects on CRC cell growth. Additionally, C8orf48 has been identified as a direct target of miR-556, further elucidating its regulatory mechanisms. The findings underscore the potential of C8orf48 as a novel target for CRC prediction and therapy.
TRUSTED_SUSTAINABILITY

Spécification

Antigène C8orf48
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugué Unconjugated
Formule PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Expression C8orf48
Numéro d’ordre du gène Q96LL4
Alias de gène C8orf48; chromosome 8 open reading frame 48; uncharacterized protein C8orf48
Symboles de gène(s) C8orf48
Espèces hôtes Rabbit
Immunogène Recombinant protein corresponding to Human C8orf48. Recombinant protein control fragment (Product #RP-93867).
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 157773
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.