missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein
Human CAMK1G (CaMKI gamma), GST Tag Recombinant Protein
Marque: Invitrogen™ A30489
Informations supplémentaires : Poids : 0.10000kg
Description
- This CAMK1G (CaMKI gamma) recombinant human protein (full length) was expressed in insect cells
- CAMK1G serine/threonine kinase is a member of CAMK family and is a novel membrane-anchored neuronal Ca2+ calmodulin-dependent protein kinase
- Full activation of CAMK1G requires both Ca2+ and calmodulin and phosphorylation by CAMKK
- CAMK1G transcripts are most abundant in neurons, with the highest levels in limited nuclei such as the central nucleus of the amygdala (CeA) and the ventromedial hypothalamus
- CAMK1G (CaMKI gamma) recombinant human protein is also available in our SelectScreen™ Kinase Profiling Service
- Validated for use with the Z'-LYTE™ assay
- Available in multiple pack sizes for convenient use
Spécification
NP_065172 | |
79.8 kDa | |
CAMK1 Kinase Family | |
Human | |
GST Tag | |
Dry Ice | |
Full Length, Recombinant, Active |
Kinase Assay | |
10 μg | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Insect | |
Liquid |