Learn More
ACROBiosystems Canine BCMA / TNFRSF17 Protein, His Tag
Marque:  ACROBiosystems BCA-C52H3-100ug
Informations supplémentaires : Poids : 0.00010kg
Code produit. 17266060
  • 423.38€ / 100µg
Expédition estimée: 30-04-2024
Description
Tumor necrosis factor receptor superfamily member 17 (TNFRSF17) is also known as B-cell maturation protein (BCMA), CD antigen CD269, which is a member of the TNF-receptor superfamily. TNFRSF17 contains one TNFR-Cys repeat. TNFRSF17 is expressed in mature B-cells, but not in T-cells or monocytes. TNFRSF17 is receptor for TNFSF13B/BLyS/BAFF and TNFSF13/APRIL. TNFRSF17 promotes B-cell survival and plays a role in the regulation of humoral immunity. TNFRSF17 can activate NF-kappa-B and JNK.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 7.5 kDa.
Spécification
Spécification
7.5 kDa
Canine BCMA, His Tag (BCA-C52H3) is expressed from human 293 cells (HEK293). It contains AA Met 1 - Asn 44 (Accession # J9NXG8-1).
-20°C
BCMA
Canine
HEK293
Bio-Markers & CD Antigens
100 μg
Met 1 - Asn 44
TNFRSF17,CD269,BCM,BCMA
Unconjugated
His Tag
Powder