Learn More
Thermo Scientific™ CaptureSelect™ Biotin Anti-AAV8 Conjugate
CaptureSelect™ Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8).
514.00€ - 1802.00€
Spécification
Contenu et stockage | Store in freezer (-5°C to -30°C). |
---|---|
Gamme de produits | CaptureSelect™ |
Type de ligand | Antibody |
Conditions d’expédition | Dry Ice |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
18416398
|
Thermo Scientific™
7103382100 |
100 μg |
514.00€
100µg |
En stock Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
18416638
|
Thermo Scientific™
7103382500 |
500 μg |
1802.00€
500µg |
En stock Connectez-vous pour voir le stock disponible |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | ||||
Description
CaptureSelect™ Biotin Anti-AAV8 Conjugate consists of a 13 kDa recombinant single domain antibody fragment (VHH affinity ligand) that binds with high affinity to adeno-associated virus particles of serotype 8 (AAV8). The affinity ligand is chemically conjugated to biotin via an appropriate spacer that retains the binding reactivity of the ligand when used in combination with streptavidin-based conjugates or streptavidin pre-coated surfaces. This conjugate allows you to detect, quantitate, and characterize AAV8 particles.
Applications for CaptureSelect Biotin Anti-AAV8 Conjugate include ELISA, Gyrolab™-based immunoassays, and label-free detection platforms such as those based on surface plasmon resonance (Biacore™ and IBIS-MX96 systems) and bio-layer interferometry (ForteBio™ Octet™ systems).
Animal origin statement
CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).
Spécification
Store in freezer (-5°C to -30°C). | |
CaptureSelect™ | |
Antibody | |
Dry Ice |