Learn More
Abnova™ CARD4 (Human) Recombinant Protein
Human CARD4 partial ORF ( AAH40339, 1 a.a. - 93 a.a.) recombinant protein with GST-tag at N-terminal.
Marque: Abnova™ H00010392-Q02.10ug
Informations supplémentaires : Poids : 0.02000kg
Description
- Sequence: MEEQGHSEMEIIPSESHPHIQLLKSNRELLVTHIRNTQCLVDNLLKNDYFSAEDAEIVCACPTQPDKVRKILDLVQSKGEEVSEFFLYLLQQL
Spécification
AAH40339 | |
nucleotide-binding oligomerization domain containing 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
NOD1 | |
GST |
10392 | |
Wheat germ expression system | |
10 μg | |
CARD4, CLR7.1, NLRC1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |