missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCDC22 Monoclonal Antibody (OTI5C1)

Description
CCDC22 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
CCDC22 is a recently identified coiled-coil domain-containing protein that has been shown to bind copines, which are calcium-dependent, membrane-binding proteins that may function in calcium signaling. In rat, CCDC22 has been observed to localize in multiple regions of the brain, including the prefrontal and somatosensory cortex, dentate gyrus and thalamus, and in the ipsilateral motor neurons of the spinal cord after sciatic nerve transection suggesting that it may play a role in neuronal injury response. The human CCDC22 gene has been identified as a novel candidate gene for syndromic X-linked intellectual disability (XLID).
Spécification
Spécification
| Antigène | CCDC22 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI5C1 |
| Concentration | 1 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Expression | Ccdc22 |
| Numéro d’ordre du gène | O60826 |
| Alias de gène | AI481216; Ccdc22; ccdc22 {ECO:0000250; coiled-coil domain containing 22; coiled-coil domain-containing protein 22; coiled-coil domain-containing protein 22 {ECO:0000250; CXorf37; DXImx40e; JM1; Ppp1r3f; RGD1560910; RTSC2; Sfc22; UniProtKB:O60826} |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?