missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117 (c-Kit) Mouse, anti-Human, APC, Clone: 104D2, STEMCELL Technologies
Description
- The 104D2 antibody reacts with CD117 (c-Kit), an ∼145 kDa type 1 transmembrane receptor for c-Kit ligand (stem cell factor/steel factor) that is broadly expressed on hematopoietic stem cells in bone marrow, including pluripotent and erythroid progenitor cells and B and T lymphocyte precursors, as well as on mast cells and melanocytes
- CD117 is expressed on ∼1 - 4% of bone marrow cells, the majority of which (∼50 - 70%) co-express CD34 and comprise progenitor cells and their precursors
- CD117 belongs to the tyrosine kinase receptor family and possesses five immunoglobulin-like C2-type domains and a cytoplasmic protein kinase domain
- Binding of c-Kit ligand to CD117 induces dimerization and autophosphorylation, which activates several intracellular signaling pathways critical for the proliferation and differentiation of hematopoietic stem cells
- Binding of the 104D2 antibody does not block binding of c-Kit ligand or antibody clone SR1
Spécification
Spécification
| Antigène | CD117 (c-Kit) |
| Applications | CyTOF, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | 104D2 |
| Conjugué | APC |
| Espèces hôtes | Mouse |
| Quantité | 25 Tests |
| Primaire ou secondaire | Primary |
| Espèces cibles | Cynomolgus, Human |
| Isotype | IgG1 κ |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?