Learn More
CD11c (Dendritic Cell Marker) Mouse, Clone: ITGAX/2507, enQuireBio™
Code produit 15920298
Modifier l'affichage
Click to view available options
Quantité:
100 μg
20 μg
Conditionnement:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15920298 100 μg 100µg
15910298 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15920298 Fournisseur EnquireBio Code fournisseur 3687MSM6P1
Please to purchase this item. Need a web account? Register with us today!
Mouse Monoclonal Antibody
Spécification
Antigène CD11c (Dendritic Cell Marker)
Applications Immunohistochemistry
Localisation cellulaire Cell Surface
Classification Monoclonal
Clone ITGAX/2507
Concentration 200μg/mL
Conjugué Unconjugated
Formule 10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide.
Espèces hôtes Mouse
Immunogène Recombinant fragment of human ITGAX protein (apprx. aa 637-827)
Poids moléculaire de l’antigène 145kDa
Contrôle positif THP-1 cells. Tonsil or Lymph Node
Méthode de purification Protein A/G purified from Bioreactor concentrate.
Quantité 100 μg
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 3687
Contenu et stockage Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Isotype IgG1 κ