Learn More
CD142 Mouse anti-Human, PE, Clone: HTF-1, BD
Description
Reacts with a 45-47 kDa, single chain, type I transmembrane protein also known as Tissue Factor (TF). CD142 has been referred in the literature as coagulation Factor III or thromboplastin and it is expressed on activated endothelial cells and lipopolysaccharide (LPS)-stimulated monocytes/macrophages. TF associates with factor VIIa to form a complex and acts as an enzyme that initiates the blood coagulation cascade. It is known as the major initiator of clotting in normal hemostasis. CD142 can be induced by various inflammatory mediators in monocytes and vascular endothelial cells. This antibody is useful in inflammation, thrombosis and hemostasis research.
Spécification
Spécification
| Antigène | CD142 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HTF-1 |
| Conjugué | PE |
| Formule | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Espèces hôtes | Mouse |
| Méthode de purification | Affinity Purified |
| Quantité | 25 Tests |
| État réglementaire | RUO |
| Afficher plus |
For Research Use Only.
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.