missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD18 integrin beta 2 subunit is a 90 kDa type I transmembrane protein expressed on all leucocytes. CD18 can combine with integrin molecules CD11a-c to form heterodimers at the cell surface, and these heterodimers are known to participate in the process of cell adhesion as well as cell-surface mediated signaling. CD18 forms heterodimers with four types of CD11 molecule to constitute leukocyte (beta2) integrins: alphaLbeta2 (CD11a/CD18, LFA-1), alphaMbeta2 (CD11b/CD18, Mac-1, CR3), alphaXbeta2 (CD11c/CD18) and alphaDbeta2 (CD11d/CD18). In most cases, the response mediated by the integrin is a composite of the functions of its individual subunits, and these integrins are essential for proper leukocyte migration, mediating intercellular contacts. Absence of CD18 leads to leukocyte adhesion deficiency-1, severe reduction of CD18 expression leads to the development of a psoriasiform skin disease. CD18 is also a target of Mannheimia (Pasteurella) haemolytica leukotoxin and is sufficient to mediate leukotoxin-mediated cytolysis. Defects in the CD18 gene are the cause of leukocyte adhesion deficiency type I (LAD1). Two transcript variants encoding the CD18 protein have been identified.
Spécification
Spécification
| Antigène | CD18 |
| Applications | Immunohistochemistry (Paraffin), Flow Cytometry, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.31 mg/mL |
| Conjugué | Unconjugated |
| Formule | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| Expression | ITGB2 |
| Numéro d’ordre du gène | P05107, P11835 |
| Alias de gène | CD18, Integrin beta 2, ITGB2, LAD, LCAMB, LFA 1, MAC 1, MF17, MFI7 |
| Symboles de gène(s) | ITGB2 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?