Learn More
Invitrogen™ CD221 (IGF1R) Monoclonal Antibody (1H7), Alexa Fluor™ 488, eBioscience™

Description
Description: This 1H7 monoclonal antibody reacts with human insulin-like growth factor-1 receptor (IGF-1R), also known as CD221. This receptor is a cell surface-expressed glycoprotein composed of two extracellular alpha subunits and two transmembrane beta subunits that possess tyrosine kinase activity. Expressed on nearly all cell types, this receptor binds IGF-I and IGF-II, as well as insulin. Ligand binding leads to activation of the PI3K/Akt and MAPK pathways, which mediate cell proliferation and survival. Many tumors and transformed cells display altered IGF-1R expression. This monoclonal antibody has been reported to block binding of IGF-I and IGF-II to the IGF-1R. Applications Reported: This 1H7 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 1H7 antibody has been pre-titrated and tested by flow cytometric analysis of the HeLa cell line. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser. Filtration: 0.2 μm post-manufacturing filtered.
Spécification
Spécification
| Antigène | CD221 (IGF1R) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 1H7 |
| Concentration | 5 μL/Test |
| Conjugué | Alexa Fluor 488 |
| Formule | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Expression | IGF1R |
| Numéro d’ordre du gène | P08069 |
| Alias de gène | A330103N21Rik; CD221; CD221 antigen; D930020L01; hyft; IGF 1 receptor; IGF 1R; IGF1R; IGF-1R; IGF-I receptor; IGFIR; IGFIRC; IGFR; Igfr1; insulin like growth factor 1 receptor; Insulin like growth factor 1 receptor precursor; insulin-like growth factor 1 receptor; Insulin-like growth factor 1 receptor alpha chain; Insulin-like growth factor 1 receptor beta chain; Insulin-like growth factor I receptor; JTK13; line 186; soluble IGF1R variant 1; soluble IGF1R variant 2 |
| Afficher plus |
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.