missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD223 (LAG-3) Monoclonal Antibody (eBioC9B7W (C9B7W)), Brilliant Violet™ 480, eBioscience™, Invitrogen™

Description
CD223 (LAG-3) Monoclonal antibody specifically detects CD223 (LAG-3) in Mouse samples. It is validated for Flow Cytometry
LAG-3 is a 70-kDa surface glycoprotein in the immunoglobulin superfamily, with structural homology to CD4. It binds to MHC class II molecules with higher affinity than CD4 and is involved in the negative regulation of T cell activation and homeostatic proliferation. LAG-3 is expressed on the surface of activated T cells, including regulatory T cells, and NK cells. Notably, CD8+ T cells express LAG-3 at significantly higher levels than CD4+ T cells. Additionally, the coexpression of LAG-3 and CD49b has been proposed as a marker for identifying Type 1 regulatory T cells (Tr1 cells) in both humans and mice. This highlights LAG-3s role in immune regulation and its potential as a target for therapeutic interventions in immune-related conditions.
Spécification
Spécification
| Antigène | CD223 (LAG-3) |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | eBioC9B7W (C9B7W) |
| Concentration | 0.2 mg/mL |
| Conjugué | Brilliant Violet 480 |
| Formule | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Expression | LAG3 |
| Numéro d’ordre du gène | Q61790 |
| Alias de gène | Activation-induced cytidine deaminase-linked autoimmunity protein; Aida; CD223; FDC; LAG3; LAG-3; Ly66; lymphocyte activating 3; lymphocyte activation gene 3 protein; lymphocyte-activation gene 3; Secreted lymphocyte activation gene 3 protein; sLAG 3; sLAG3; sLAG-3; soluble LAG 3lymphocyte activating 3; soluble LAG3 |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?