missing translation for 'onlineSavingsMsg'
Learn More

CD294 (CRTH2) (extracellular) Polyclonal Antibody, Invitrogen™

Code produit 17129803
Change view
Click to view available options
Quantité:
50 μL
Conditionnement:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17129803 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17129803 Fournisseur Invitrogen Code fournisseur PA5111888

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Cet article est supprimé, vous trouverez le remplacement et les alternatives dans les onglets respectifs ci-dessous s'ils ont été identifiés
Voir les produits de remplacement

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Reconstitute antibody with 50 μL of double distilled water (DDW). The reconstituted solution can be stored at 4 degrees C for up to 1 week. For longer periods, small aliquots should be stored at -20 degrees C. Avoid multiple freezing and thawing. Centrifuge all antibody preparations before use (10000 x g 5 min). Peptide sequence is: (C)RAHSVTT LRQLASR Homology: Mouse amino acid residues are identical. Will not recognize DP2 receptor from human samples.

CRTH2 (GPR44) is a chemoattractant receptor-homologous molecule expressed on Th2 cells that is an identified receptor for the prostaglandin D2 (PGD2) and prostaglandin D receptor. CRTH2 is expressed on Th2 cells and eosinophils and mediates chemotaxis of these cells to PGD2 and is thus thought to be a key receptor mediating eosinophil and Th2 recruitment during allergic responses. However, CRTH2-null mice showed enhanced eosinophil recruitment into the lung consistent with observations that the CRTH2-null mice produced significantly higher amounts of interleukin-5 (IL-5) and IL-3. This suggests that CRTH2 plays a nonredundant role in restricting eosinophilia and allergic response in vivo. CRTH2 is also a novel member of the G protein-coupled leukocyte chemoattractant receptor family, which is selectively expressed in Th2 but not Th1 lineage cells. CRTH2 has been reported to be expressed in the central nervous system, peripheral tissues, and in blood cells, particularly activated T helper-2 cells. Two transcripts have been isolated: a 3.5 kb transcript expressed primarily in the thalamus, frontal cortex, pons and hippocampus; and a 3.4 kb transcript expressed in fetal liver, leukocytes and thymus. CRTH2 is selectively expressed in vivo in an activated state of Th2 cells including allergen-responsive Th2 cells, suggesting its pivotal roles in ongoing Th2-type immune reactions. At least two different isoforms of CRTH2 are known to exist. Single nucleotide polymorphisms in the 3′ UTR of the CRTH2 gene have been associated with asthma susceptibility.
TRUSTED_SUSTAINABILITY

Spécification

Antigène CD294 (CRTH2) (extracellular)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.8 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA and 0.05% sodium azide; pH 7.4
Expression PTGDR2
Numéro d’ordre du gène Q6XKD3, Q9Z2J6
Alias de gène CD294; Chemoattractant receptor-homologous molecule expressed on Th2 cells; CRTH2; DL1R; DP2; G protein-coupled receptor 44; GPR44; G-protein coupled receptor 44; Grp45; MGC130436; Prostaglandin D2 receptor 2; PTGDR2
Symboles de gène(s) PTGDR2
Espèces hôtes Rabbit
Immunogène Peptide corresponding to amino acid residues 270-283 of rat PTGDR2 (3rd extracellular loop)
Méthode de purification Antigen affinity chromatography
Quantité 50 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 14764, 309212
Espèces cibles Mouse, Rat
Contenu et stockage -20°C
Type de produit Antibody
Forme Lyophilized
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.