missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD299 (DC-SIGN/L) Polyclonal Antibody
GREENER_CHOICE

Code produit 17721664
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17721664 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17721664 Fournisseur Invitrogen™ Code fournisseur PA5120262

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Positive test controls include: HepG2. The target is usually found in the following locations: Cell membrane, Secreted, Single-pass type II membrane protein. Immunogen sequence: QVSKVPSSLS QEQSEQDAIY QNLTQLKAAV GELSEKSKLQ EIYQELTQLK AAVGELPEKS KLQEIYQELT RLKAAVGELP EKSKLQEIYQ ELTRLKAAVG ELPEKSKLQE IYQELTRLKA AVGELPEKSK LQEIYQELTE LKAAVGELPE KSKLQEIYQE LTQLKAAVGE LPDQSKQQQI YQELTDLKTA FERLCRHCPK DWTFFQGNCY FMSNSQRNWH DSVTACQEVR AQLVVIKTAE EQNFLQLQTS RSNRFSWMGL SDLNQEGTWQ WVDGSPLSPS FQRYWNSGEP NNSGNEDCAE FSGSGWNDNR CDVDNYWICK KPAACFRDE.

CLEC4M is a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells.Western blots using two different antibodies against two unique regions of this protein target confirm the same apparent molecular weight in our tests.This gene encodes a type II integral membrane protein that is 77% identical to CD209 antigen, a HIV gp120-binding protein. This protein, like CD209, efficiently binds both intercellular adhesion molecule 3 (ICAM3) and HIV-1 gp120, and enhances HIV-1 infection of T cells. This gene is mapped to 19p13.3, in a cluster with the CD209 and CD23/FCER2 genes. Multiple alternatively spliced transcript variants have been found for this gene, but the biological validity of some variants has not been determined.
TRUSTED_SUSTAINABILITY

Spécification

Antigène CD299 (DC-SIGN/L)
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.065 mg/mL
Conjugué Unconjugated
Formule PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Expression CLEC4M
Numéro d’ordre du gène Q9H2X3
Alias de gène CD209 antigen-like protein 1; Cd209b; CD209b antigen; CD209L; CD209L1; CD299; CD299 antigen; CLEC4M; C-type lectin domain family 4 member M; C-type lectin domain family 4, member M; DC-SIGN2; DCSIGNR; DC-SIGNR; DC-SIGN-related protein; Dendritic cell-specific ICAM-3-grabbing non-integrin 2; HP10347; liver/lymph node-specific ICAM-3 grabbing non-integrin; liver/lymph node-specific ICAM-3-grabbing non-integrin; LSIGN; L-SIGN; mannose binding C-type lectin DC-SIGNR; MGC129964; MGC47866
Symboles de gène(s) CLEC4M
Espèces hôtes Rabbit
Immunogène A synthetic peptide corresponding to a sequence within amino acids 71-399 of human CLEC4M (NP_0550723).
Méthode de purification Affinity Chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 10332, 288378
Espèces cibles Human, Mouse, Rat
Contenu et stockage -20°C, Avoid Freeze/Thaw Cycles
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.