Learn More
CD3 Mouse anti-Human, FITC, Clone: HIT3A, BD
Code produit 15871349
Change view
Click to view available options
Quantité:
100 Tests
25 Tests
Conditionnement:
1 pièce
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15871349 25 Tests 1 pièce
15838078 100 Tests 1 pièce
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 15871349 Fournisseur BD Biosciences Code fournisseur 561802
Mouse Monoclonal Antibody
The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
TRUSTED_SUSTAINABILITY
Spécification
Antigène CD3
Applications Flow Cytometry
Classification Monoclonal
Clone HIT3A
Conjugué FITC
Formule Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Espèces hôtes Mouse
Méthode de purification Affinity Purified
Quantité 25 Tests
État réglementaire RUO
Primaire ou secondaire Primary
Espèces cibles Human
Contenu et stockage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Afficher plus Afficher moins
For Research Use Only.