missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD44var (v7-v8) Monoclonal Antibody (VFF-17), Biotin, eBioscience™
GREENER_CHOICE

Code produit 15551387
Change view
Click to view available options
Quantité:
100 Tests
Conditionnement:
100 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15551387 100 Tests 100 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15551387 Fournisseur Invitrogen™ Code fournisseur BMS118BT

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

Description: Specifically recognizes an epitope encoded by exons v7-v8 on the variant portion of human CD44. Standard CD44 (CD44s) lacks the entire variable region and is found mainly in cells of lymphohematopoietic origin. The multiple isoforms (designated CD44v; v = variant) contain alternatively spliced exons inserted within the extracellular domain. These isoforms are known as CD44v1, CD44v2, CD44v3, CD44v4, CD44v5, CD44v6, CD44v7, CD44v8, CD44v9, CD44v10, CD44v11. Applications Tested: Flow Cytometry. Purity: >95%.

CD44 cell surface antigen is a 100kDa glycoprotein widely expressed on human leucocytes, white matter of the brain and by some epithelial cells of the intestine and breast. Several isoforms of CD44 exist, including the predominant CD44H isoform detected in many normal tissues. CD44 is a receptor for hyaluronic acid (HA) and is involved in cell-cell interactions, cell adhesion and migration. CD44 also participates in a wide variety of cellular functions including lymphocyte activation, recirculation and homing. CD44 expression may be up-regulated upon some carcinomas, and it has been speculated that this may be related to metastatic potential.
TRUSTED_SUSTAINABILITY

Spécification

Antigène CD44var (v7-v8)
Applications Flow Cytometry
Classification Monoclonal
Clone VFF-17
Concentration 10 μL/Test
Conjugué Biotin
Formule TBS with 1% BSA and 0.02% sodium azide; pH 7.2-7.4
Expression CD44
Numéro d’ordre du gène P16070
Alias de gène AL133330.1; CD44; CD44 antigen; CD44 molecule (Indian blood group); CD44 splice variant; CD44 var; CD44 variant; CD44v7; CD44v7-8; CD44v7-v8; CD44v8; CDW44; cell surface glycoprotein CD44; chondroitin sulfate proteoglycan 8; CSPG8; ECMRIII; ECMR-III; Epican; extracellular matrix receptor III; GP90 lymphocyte homing/adhesion receptor; H-CAM v7; H-CAM v8; HCELL; hematopoietic cell E- and L-selectin ligand; heparan sulfate proteoglycan; Hermes antigen; homing function and Indian blood group system; HUTCHI; HUTCH-I; Hyaluronate receptor; IN; LHR; MC56; MDU2; MDU3; MIC4; PGP I; Pgp1; PGP-1; PGP-I; Phagocytic glycoprotein 1; Phagocytic glycoprotein I; soluble CD44
Symboles de gène(s) CD44
Espèces hôtes Mouse
Méthode de purification Affinity Chromatography
Quantité 100 Tests
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 960
Espèces cibles Human
Contenu et stockage 2-8°C
Type de produit Antibody
Forme Liquid
Isotype IgG2b
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.