missing translation for 'onlineSavingsMsg'
Learn More

CD45 Antibody (OX-1), Alexa Fluor™ 594, Novus Biologicals™

Code produit 18730912
Change view
Click to view available options
Quantité:
0.25 mL
Conditionnement:
250µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
18730912 0.25 mL 250µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18730912 Fournisseur Novus Biologicals (Bio-Techne) Code fournisseur NB10064895AF594

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

CD45 Monoclonal antibody specifically detects CD45 in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY

Spécification

Antigène CD45
Applications Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OX-1
Conjugué Alexa Fluor 594
Dilution Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Formule 50mM Sodium Borate
Alias de gène B220, CD_antigen: CD45, CD45, CD45 antigen, CD45R, EC 3.1.3.48, EC:3.1.3.48, GP180, LCA, L-CA, Leukocyte common antigen, LY5, protein tyrosine phosphatase, receptor type, c polypeptide, PTPRC, receptor-type tyrosine-protein phosphatase C, T200, T200 glycoprotein, T200 leukocyte common antigen
Espèces hôtes Mouse
Immunogène This CD45 Antibody (OX-1) was prepared from A 100kD thymocyte glycoprotein fraction enriched for the rat leukocyte common antigen (L-CA).
Méthode de purification Protein G purified
Quantité 0.25 mL
État réglementaire RUO
Disciplines de recherche Adaptive Immunity, Autoimmune Diseases, Cell Biology, Cellular Markers, Cellular Signaling, Cytokine Research, Glia Markers, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Inflammation, Innate Immunity, MAP Kinase Signaling, Mast Cell Markers, Mesenchymal Stem Cell Markers, Microglia Markers, Modulation of DNA Pools, Myeloid Cell Markers, Myeloid derived Suppressor Cell, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primaire ou secondaire Primary
Identification génétique (Entrez) 5788
Espèces cibles Rat
Contenu et stockage Store at 4°C in the dark.
Forme Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.