missing translation for 'onlineSavingsMsg'
Learn More

CD52 Monoclonal Antibody (CF1D12), PE-Cyanine7, eBioscience™, Invitrogen™

Code produit p-7159061
Change view
Click to view available options
Quantité:
100 Tests
25 Tests
Unit Size:
100 pièces
25 pièces
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantité unitSize
16390744 25 Tests 25 pièces
16300754 100 Tests 100 pièces
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16390744 Supplier eBioscience Supplier No. 25052941

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

This CF1D12 monoclonal antibody reacts with human CD52, a cell surface glycoprotein expressed in the immune and reproductive systems. CF1D12 is thought to recognize the carbohydrate region of CD52, not the protein core of the molecule. Tissue-specific mod

Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 775 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser Human CD52, also known as Cambridge pathology antigen 1 (CAMPATH-1), is a glycoprotein found in the male reproductive tract and immune cells. It is abundantly expressed on the surface of nearly all human peripheral blood lymphocytes. The CD52 antigen is GPI-anchored membrane receptor that consists of a small 12 amino acid peptide attached to a large N-linked carbohydrate group. CD52 is a potent target for inducing cytolysis of mature lymphocytes. Alemtuzumab, a humanized monoclonal antibody targeting CD52, is used as an immunomodulatory drug for the treatment of chronic lymphocytic leukemia, T cell lymphomas, multiple sclerosis, and other diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigène CD52
Applications Flow Cytometry
Classification Monoclonal
Clone CF1D12
Concentration 5 μL/Test
Conjugué PE-Cyanine7
Formule PBS with 0.1% gelatin, 0.2% BSA and 0.09% sodium azide; pH 7.2
Expression CD52
Numéro d’ordre du gène P31358
Alias de gène Cambridge pathology 1 antigen; CAMPATH-1 antigen; CDW52; Epididymal secretory protein E5; epididymis secretory sperm binding protein Li 171mP; He5; HEL-S-171mP; Human epididymis-specific protein 5
Symboles de gène(s) CD52
Espèces hôtes Mouse
Immunogène Human CD52
Méthode de purification Affinity Chromatography
Quantité 25 Tests
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1043
Espèces cibles Human
Contenu et stockage 4° C, store in dark, DO NOT FREEZE!
Type de produit Antibody
Forme Liquid
Isotype IgG3 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.