missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CDYL Polyclonal Antibody

Description
Recommended positive controls: NT2D1, U87-MG, mouse kidney. Predicted reactivity: Mouse (84%), Rat (84%), Bovine (88%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Chromodomain Y is a primate-specific Y-chromosomal gene family expressed exclusively in the testis and implicated in infertility. Although the Y-linked genes are testis-specific, this autosomal gene is ubiquitously expressed. The Y-linked genes arose by retrotransposition of an mRNA from this gene, followed by amplification of the retroposed gene. Proteins encoded by this gene superfamily possess a chromodomain, a motif implicated in chromatin binding and gene suppression, and a catalytic domain believed to be involved in histone acetylation. Multiple proteins are encoded by transcript variants of this gene.
Spécification
Spécification
| Antigène | CDYL |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.54 mg/mL |
| Conjugué | Unconjugated |
| Formule | 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 |
| Expression | CDYL |
| Numéro d’ordre du gène | Q9WTK2, Q9Y232 |
| Alias de gène | AI325931; CDYL; CDYL1; CDY-like; CDY-like, autosomal; chromodomain protein Y-like variant 2; chromodomain protein Y-like variant 3; chromodomain protein Y-like variant 4; chromodomain protein, Y chromosome-like; chromodomain protein, Y-like; chromodomain Y like; chromodomain Y-like protein; chromodomain Y-like protein pseudogene; Crotonyl-CoA hydratase; DKFZp586C1622; MGC131936; Putative histone acetyltransferase Cdyl; Putative tubulin acetyltransferase Cdyl; testis-specific chromodomain Y-like protein |
| Symboles de gène(s) | CDYL |
| Afficher plus |
Nom du produit
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?