Learn More
Chlorure de césium, 99 % (base métallique), Thermo Scientific Chemicals
Description
Evacuation of radio tubes, manufacturing of cesium compounds, brewing, x-ray fluorescent screens, radiographic contrast medium.Cesium chloride is widely utilized as a precursor for the source of cesium ions. It finds application in analytical chemistry and acts as a reagent to identify inorganic ions like aluminum, gallium and chromium by the color and morphology of the precipitate. Its radioisotopes are employed in nuclear medicine applications such as treatment of cancer and diagnosis of myocardial infarction. Occasionally, it can act as a phase transfer catalyst in selected organic reactions. It is applied in the manufacturing of electrically conducting glasses and screens of cathode ray tubes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Évacuation de tubes radio, fabrication de composés de césium, brassage, écrans fluorescents à rayons X, milieu de contraste radiographique
Spécification
Spécification
| Point d’ébullition | 1303°C |
| Nom chimique ou matériau | Cesium chloride |
| Point de fusion | 646°C |
| Forme physique | Cristalline |
| Quantité | 50 g |
| Plage de pourcentage du dosage | 99% (Metals basis) |
| Notes de pourcentage du dosage | (Base des métaux) |
| Merck Index | 14,2011 |
| Informations sur la solubilité | Very soluble in water; Soluble in alcohol; Insoluble in acetone. |
| Nom IUPAC | chlorure de césium |
| Afficher plus |
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.