missing translation for 'onlineSavingsMsg'
Learn More

CIB1 Monoclonal Antibody (OTI1D7), TrueMAB™, OriGene

Code produit 17524874
Encompass_Preferred
Change view
Click to view available options
Quantité:
30 μL
100 μL
Conditionnement:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
17524874 30 μL 30µL
17504874 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 17524874 Fournisseur Origene Technologies Code fournisseur TA811149S

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Mouse Monoclonal Antibody

CIB1 Monoclonal antibody specifically detects CIB1 in Human samples. It is validated for Western Blot

Specifications

Antigène CIB1
Applications Western Blot
Classification Monoclonal
Clone OTI1D7
Concentration 1 mg/mL
Conjugué Unconjugated
Formule PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Expression SF3B6
Numéro d’ordre du gène Q9Y3B4
Alias de gène calcium and integrin binding 1; calcium and integrin binding 1 (calmyrin); calcium and integrin binding protein 1; calcium and integrin binding, protein kinase interacting protein; Calcium- and integrin-binding protein; calcium and integrin-binding protein 1; calmyrin; CIB; CIB1; Cibkip; CIBP; DNA-dependent protein kinase interacting protein; DNA-PK interaction protein; DNA-PKcs-interacting protein; Kinase-interacting protein; KIP; KIP1; LOW QUALITY PROTEIN: calcium and integrin-binding protein 1; PRKDCIP; protein kinase, DNA activated, catalytic polypeptide interacting protein; Sip2-28; SNK-interacting protein 2-28; testicular secretory protein Li 9
Espèces hôtes Mouse
Immunogène Full length human recombinant protein of human SF3B14 produced in E.coli.
Méthode de purification Affinity chromatography
Quantité 30 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 51639
Espèces cibles Human
Contenu et stockage -20° C, Avoid Freeze/Thaw Cycles
Forme Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.