missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Codeine N-Oxide, Mikromol™
missing translation for 'orderingAttributeHoverText'
Quantité:
250 mg
missing translation for 'unitSize'
250mg
Description
Codeine N-Oxide
Specifications
Specifications
| Nom chimique ou matériau | Codeine N-Oxide |
| CAS | 3688-65-1 |
| Noms d’analytes ou de composants | Codeine N-Oxide |
| Conditions d’expédition | Room Temperature |
| Formule moléculaire | C18 H21 N O4 |
| Certifications/Conformité | ISO 17025 |
| Synonyme | Codeine N-Oxide, Codeine N-oxide (6CI,7CI), Codeigene, Gencodeine, Genkodein, Genocodein, Genocodeine, Morphinan-6-ol, 7,8-didehydro-4,5-epoxy-3-methoxy-17-methyl-, 17-oxide, (5alpha,6alpha)-, Morphinan-6alpha-ol, 7,8-didehydro-4,5alpha-epoxy-3-methoxy-17-methyl-, 17-oxide (8CI) |
| Formule InChI | InChI=1S/C18H21NO4/c1-19(21)8-7-18-11-4-5-13(20)17(18)23-16-14(22-2)6-3-10(15(16)18)9-12(11)19/h3-6,11-13,17,20H,7-9H2,1-2H3/t11-,12+,13-,17-,18-,19?/m0/s1 |
| SMILES | COc1ccc2C[C@@H]3[C@@H]4C=C[C@H](O)[C@@H]5Oc1c2[C@]45CC[N+]3(C)[O-] |
| Nom IUPAC | (4R,4aR,7S,7aR,12bS)-9-methoxy-3-methyl-3-oxido-2,4,4a,7,7a,13-hexahydro-1H-4,12-methanobenzofuro[3,2-e]isoquinoline-3-ium-7-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction