missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ COL17A1 Polyclonal Antibody
GREENER_CHOICE

Code produit 15740502 Tous les produits Thermo Scientific Produits
Change view
Click to view available options
Quantité:
100 μL
Conditionnement:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
15740502 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 15740502 Fournisseur Invitrogen™ Code fournisseur PA562710

Veuillez vous pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

Rabbit Polyclonal Antibody

Immunogen sequence: NLPSHVWSST LPAGSSMGTY HNNMTTQSSS LLNTNAYSAG SVFGVPNNMA SCSPTLHPGL STSSSVFGMQ NNLAPSLTTL SHGTTTTSTA Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 84%.

COL17A1 is the alpha chain of type XVII collagen. Unlike most collagens, collagen XVII is a transmembrane protein, and a structural component of hemidesmosomes, multiprotein complexes at the dermal-epidermal basement membrane zone that mediate adhesion of keratinocytes to the underlying membrane. COL17A1 is a member of the FACIT collagen family (fibril-associated collagens with interrupted helices). Moreover, COL17A1 is localized to tissues containing type I collagen so, like other members of this collagen family, it may serve to maintain the integrity of the extracellular matrix. Diseases associated with COL17A1 dysfunction include epithelial recurrent erosion dystrophy and epidermolysis bullosa.
TRUSTED_SUSTAINABILITY

Spécification

Antigène COL17A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugué Unconjugated
Formule PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Expression COL17A1
Numéro d’ordre du gène Q9UMD9
Alias de gène 120 kDa linear IgA dermatosis antigen; 120 kDa linear IgA disease antigen; 120 kDa linear IgA disease antigen homolog; 180 kDa bullous pemphigoid antigen 2; 97 kDa LAD antigen; 97 kDa linear IgA bullous dermatosis antigen; 97 kDa linear IgA disease antigen; alpha 1 type XVII collagen; BA16H23.2; bA16H23.2 (collagen, type XVII, alpha 1 (BP180)); BP180; BPA-2; Bpag; BPAG2; Bullous pemphigoid antigen 2; bullous pemphigoid antigen 2 (180kD); Col17a1; Collagen alpha-1(XVII) chain; collagen type XVII alpha 1; collagen XVII, alpha-1 polypeptide; collagen, type XVII, alpha 1; ERED; LABD97; LAD-1; Linear IgA bullous disease antigen of 97 kDa; Linear IgA disease antigen 1; procollagen, type XVII, alpha 1; type XVII collagen alpha-1
Symboles de gène(s) COL17A1
Espèces hôtes Rabbit
Immunogène Recombinant protein corresponding to Human COL17A1. Recombinant protein control fragment (Product #RP-103143).
Méthode de purification Antigen affinity chromatography
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 1308
Espèces cibles Human
Contenu et stockage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Type de produit Antibody
Forme Liquid
Isotype IgG
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.