Learn More
CoraLite™ Plus 488-conjugated CD31 Mouse Monoclonal Antibody, Proteintech
Code produit 30235713 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
20 μL
Conditionnement:
20µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30235713 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Code produit 30235713 Fournisseur Proteintech Code fournisseur CL4886606520UL
Mouse Monoclonal Antibody
The CD31 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD31. This antibody recognizes human antigen. The CD31 antibody has been validated for the following applications: IF/ICC, IF-P analysis. This antibody is coralite™ plus 488.
Spécification
Antigène CD31
Applications Immunocytochemistry/Immunofluorescence, Immunofluorescence
Classification Monoclonal
Clone 3F8E2
Concentration 1000 μg/mL
Conjugué CoraLite Plus 488
Symboles de gène(s) CD31; FLJ58394; PECAM-1
Espèces hôtes Mouse
Immunogène Recombinant Protein
Méthode de purification Protein A purification
Quantité 20 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 5175
Espèces cibles Human
Contenu et stockage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Type de produit Conjugated Antibody
Forme Liquid
Isotype IgG1