Learn More
CoraLite™ Plus 488-conjugated DLC1 Mouse Monoclonal Antibody, Proteintech
Code produit 30233319 Tous les produits Proteintech Produits
Change view
Click to view available options
Quantité:
100 μL
20 μL
Conditionnement:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité unitSize
30233319 100 μL 100µL
30233961 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Code produit 30233319 Fournisseur Proteintech Code fournisseur CL48866894100UL
Mouse Monoclonal Antibody
The DLC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human DLC1. This antibody recognizes human antigen. The DLC1 antibody has been validated for the following applications: IF/ICC analysis. This antibody is coralite™ plus 488.
Spécification
Antigène DLC1
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 1G10H12
Concentration 1000 μg/mL
Conjugué CoraLite Plus 488
Symboles de gène(s) ARHGAP7; FLJ21120; HP; STARD12; p122-RhoGAP
Espèces hôtes Mouse
Immunogène Recombinant Protein
Méthode de purification Protein G purification
Quantité 100 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 10395
Espèces cibles Human
Contenu et stockage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Poids moléculaire des anticorps 115 kDa
Type de produit Conjugated Antibody
Forme Liquid
Isotype IgG1